missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ Double-Stranded cDNA Synthesis Kit

Product Code. 10729803
Change view
Click to view available options
Quantity:
10 Reactions
50 Reactions
Unit Size:
10 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10729803 10 Reactions 10 reactions
10073372 50 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10729803 Supplier Invitrogen™ Supplier No. 11917010
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Reagents and buffers for 10 first- and second-strand cDNA synthesis reactions converting 25-50μg total RNA or 0.2-5μg mRNA into double-stranded cDNA.

Requires

Primer

Maximize the yield of full-length cDNA, as well as that of overall cDNA synthesis

Contains all of the reagents (except an oligo (dT)-containing primer) necessary to make high-quality, double-stranded cDNA from total RNA or poly A+-selected RNA (mRNA).

Simple, reliable, and trouble-free
The SuperScript Double-Stranded cDNA Synthesis Kit offers a simple, reliable, and trouble-free method by which RNA templates can be converted to cDNA for use in cloning and library construction experiments. The reactions described in the SuperScript Double-Stranded cDNA Synthesis Kit protocol are designed to convert 25–50μg total RNA, or 0.2–5μg mRNA, into first- and second-strand cDNA (supply your own primer). Enjoy the benefits of a tested protocol and reagents of consistent quality. To get the best possible cDNA product using this kit, use a high-quality RNA substrate.

  • Reliability: each buffer, reagent, and enzyme in the kit is of the same high quality you expect from Life Technologies
  • Performance: SuperScript II is an enzyme you can trust for high-yield, high-quality, general-purpose, first-strand synthesis
  • Optimal temperature: 42°C

DNA and RNA Purification and Analysis, DNA Labeling, Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, Nucleic Acid Labeling and Oligo Synthesis, PCR and Real-Time PCR, Reverse Transcription

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contents: The kit includes reagents and buffers for 10 first- and second-strand cDNA synthesis reactions converting 25-50 μg total RNA or 0.2-5 μg mRNA into double-stranded cDNA.

Storage: Store at -20°C. All components are guaranteed stable for 6 months when properly stored.
Format Kit
Labeling Method Direct Labeling
Optimal Reaction Temperature 42°C
Product Line SuperScript
Product Type cDNA Synthesis Kit
Quantity 10 Reactions
Reverse Transcriptase SuperScript™ II
Shipping Condition Wet Ice
For Use With (Application) Microarray Analysis
Final Product Type cDNA (Double-Stranded)
Sample Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.