missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ Indirect cDNA Labeling System

Product Code. 10073162
Change view
Click to view available options
Quantity:
30 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10073162 30 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10073162 Supplier Invitrogen™ Supplier No. L101402
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

The SuperScript™ Indirect cDNA Labeling Module includes SuperScript™ III Reverse Transcriptase, oligo(dT)20- VN (2.5μg/μL), random primers, control RNA ladder (0.5g/μL), 5X first-strand buffer, 0.1M DTT, 10mM dNTP mixture, RNaseOUT™ Recombinant Ribonuclease Inhibitor (40U/μL), DEPC-treated water, DMSO (Labeling Grade), 2X coupling buffer, 3M NaAc, pH 5.2, and glycogen (20mg/mL). The SuperScript™ Indirect cDNA Labeling System includes all of the components listed for the Labeling Module, plus S.N.A.P.™ column, collection tubes, loading buffer, wash buffer, amber collection tubes.

An array labeling kit based on proven methods of indirect cDNA labeling

In this indirect labeling method, mRNA or total RNA is reverse transcribed using SuperScript III RT, incorporating amino-modified dUTP and dATP into the synthesized cDNA. The template RNA is then degraded by base hydrolysis, and the reaction is neutralized with acid. The amino-modified cDNA is then purified to remove unincorporated nucleotides, primers, and buffers. In the second step, the modified cDNA is coupled with the active form of a fluorescent dye. The fluorescently labeled cDNA is purified with a S.N.A.P.™ spin column to remove any unreacted dye. The resulting fluorescently labeled cDNA is ready for hybridization to microarrays.

The SuperScript Indirect cDNA Labeling System provides a complete system that includes both the core kit reagents (except dyes) and a purification module. (The SuperScript Indirect cDNA Labeling Module includes just the core kit reagents. Both provide the flexibility to use fluorescent nucleotides of your choice.)

  • SuperScript III RT for generating high cDNA yields
  • A proprietary nucleotide mixture to increase signal intensity
  • A convenient kit format that saves valuable time

DNA and RNA Purification and Analysis, DNA Labeling, Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, Nucleic Acid Labeling and Oligo Synthesis

TRUSTED_SUSTAINABILITY

Specifications

Product Type cDNA Labeling System
Detection Method Fluorescence
Content And Storage The SuperScript™ Indirect cDNA Labeling Module includes SuperScript™ III Reverse Transcriptase, oligo(dT)20- VN (2.5 μg/μl), random primers, control RNA ladder (0.5 g/μl), 5X first-strand buffer, 0.1 M DTT, 10 mM dNTP mixture, RNaseOUT™ Recombinant Ribonuclease Inhibitor (40 U/μl), DEPC-treated water, DMSO (Labeling Grade), 2X coupling buffer, 3M NaAc, pH 5.2, and glycogen (20 mg/ml). The SuperScript™ Indirect cDNA Labeling System includes all of the components listed for the Labeling Module, plus S.N.A.P.™ column, collection tubes, loading buffer, wash buffer, amber collection tubes. Store all Labeling Module components at -20°C. Store S.N.A.P.™ columns, collection tubes, loading buffer, wash buffer, and amber collection tubes at room temperature. Guaranteed stable for 6 months when properly stored.
Final Product Type cDNA (Labeled)
Label or Dye Alexa Fluor™ 555, Alexa Fluor™ 647, Cy™3, Cy™5
Includes Label or Dye No
Labeling Target cDNA
Labeling Method Indirect Labeling
Sample Type RNA
Product Line SuperScript
Quantity 30 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.