missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ One-Cycle cDNA Kit for use with Thermo Scientific™ One-Cycle Assays

Product Code. 10648524 Shop All Thermo Scientific Products
Click to view available options
Quantity:
30 Reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10648524

Brand: Invitrogen™ A10752030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

50μM T7-Oligo(dT) Primer, 5X 1st Strand Reaction Mix, 0.1M DTT, 10mM dNTP, SuperScript™ II (200u/μL), 5X 2nd Strand Reaction Mix, E.Coli DNA Ligase, E.Coli DNA Polymerase I, RNase H, T4 DNA Polymerase, 0.5M EDTA, RNase-free Water

Conveniently contains all of the reagents required for double-stranded cDNA synthesis within the GeneChip™ workflow

The kit includes Invitrogen's proprietary RNase H reduced activity SuperScript™ II Reverse Transcriptase and a qualified oligo (dt)-containing primer to perform reactions standard to Affymetrix– protocol. Designed to consistently produce higher yields of full-length cDNA for Affymetrix One-Cycle assays using either total RNA or poly A+ selected RNA (mRNA).

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 50 μM T7-Oligo(dT) Primer, 5X 1st Strand Reaction Mix, 0.1M DTT, 10 mM dNTP, SuperScript™ II (200u⁄ul), 5X 2nd Strand Reaction Mix, E.Coli DNA Ligase, E.Coli DNA Polymerase I, RNase H, T4 DNA Polymerase, 0.5M EDTA, RNase-free Water
All components are guaranteed stable for 6 months when properly stored.
Detection Method Biotin-based
Format Kit
Label or Dye Biotin
Final Product Type cDNA (Double-stranded)
Sample Type Poly(A+) RNA, Total RNA
Labeling Target cDNA
For Use With (Application) For generating double-stranded cDNA
Includes Label or Dye No
For Use With (Equipment) Affymetrix GeneChip™
Labeling Method Direct Labeling
Product Line SuperScript
Product Type cDNA Kit
Quantity 30 Reactions
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.