missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Resolving Buffer

Product Code. 15527935
Change view
Click to view available options
Quantity:
2 x 500 mL
5 x 500 mL
Unit Size:
Pack of 2
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15527935 2 x 500 mL Pack of 2
15530445 5 x 500 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15527935 Supplier Invitrogen™ Supplier No. HC2212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank

Recommended Storage

Room Temperature

TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage 2 x 500 mL (1 L total) SureCast Resolving Buffer packs.

Store at room temperature.
Quantity 2 x 500 mL
Shipping Condition Room Temperature
Product Line SureCast
Product Type Resolving Buffer

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.