missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Resolving Buffer

Product Code. 15530445
Change view
Click to view available options
Quantity:
2 x 500 mL
5 x 500 mL
Unit Size:
Pack of 2
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15530445 5 x 500 mL Pack of 5
15527935 2 x 500 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15530445 Supplier Invitrogen™ Supplier No. HC2215
  • ONLINE EXCLUSIVE PRICE
    82.25 €
    66.62 € / Pack of 5
    Save 15.63 €
    19% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank

Recommended Storage

Room Temperature

Warranty

Up to 12 months

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 5 x 500 mL (2.5 L total) SureCast Resolving Buffer packs.

Store at room temperature.
Quantity 5 x 500 mL
Shipping Condition Room Temperature
Product Line SureCast
Product Type Resolving Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.