missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO

Product Code. 11550746
Change view
Click to view available options
Quantity:
500 μL
1 mL
20 x 50 μL
Unit Size:
100µL
1mL
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11550746 20 x 50 μL 100µL
10535203 500 μL Each
10463252 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11550746 Supplier Invitrogen™ Supplier No. S7586
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Displays bright fluorescence when bound to RNA and low background in gels. Invitrogen™ SYBR™ Green II RNA Gel Stain, 10,000X concentrate in DMSO is ideal for use with either formaldehyde/agarose or polyacrylamide gels.

SYBR Green II RNA gel stain is one of the most sensitive dyes known for detecting RNA in electrophoretic gels using laser scanners or standard UV transilluminators. It is significantly more sensitive than ethidium bromide, the most commonly used stain for detecting nucleic acids in gels.

Available in three convenient packaging options:

  • 500μL (stains 50 minigels)
  • 1mL (stains 100 minigels)
  • 20 × 50μL (each stains 5 minigels)—Avoids waste when processing fewer minigels

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Supplied as a 10,000X concentrate in DMSO

Store at -20°C, protected from light in a dessicator.
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye SYBR Green II
Quantity 20 x 50 μL
Shipping Condition Room Temperature
Target Molecule RNA
Product Type RNA Gel Stain

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.