missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYBR™ Safe DNA Gel Stain in 1X TAE
GREENER_CHOICE

Product Code. 10063352
Change view
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10063352 1 L 1 L
11560676 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10063352 Supplier Invitrogen™ Supplier No. S33111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Developed specifically for reduced mutagenicity to be less hazardous. Invitrogen™ SYBR™ Safe DNA Gel Stain in 1X TAE is safer and more sensitive than ethidium bromide for staining DNA in agarose or acrylamide gels. The premixed solution is available in 1L and 4L units.

SYBR Safe™ stain is not only less mutagenic than ethidium bromide, but SYBR Safe™ stain′s detection sensitivity is better than that of ethidium bromide. SYBR Safe™ stain comes as a premixed solution that can be used just like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe™ stain has fluorescence excitation maxima at ∼280 and ∼502nm, and an emission maximum at ∼530nm.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Supplied as a ready-to-use solution in 1X TAE

Store at room temperature in the original container.
Detection Location In-Gel Detection
Detection Method Fluorescence
Label or Dye SYBR Safe
Quantity 1 L
Shipping Condition Room Temperature
Target Molecule DNA
Product Type DNA Gel Stain

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.