missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ T7 RNA Polymerase

Product Code. 10093062
Change view
Click to view available options
Quantity:
2 x 2500 U
2500 U
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10093062 2500 U Each
11528906 2 x 2500 U Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10093062 Supplier Invitrogen™ Supplier No. 18033019
Focus on Regenerative Therapies
Use promo code 28027
Afficher toutes les promotions
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Includes

T7 RNA Polymerase is supplied with a vial of 5X reaction buffer [200mM Tris-HCl (pH 8.0), 40mM MgCl2 , 10mM spermidine-(HCl)3 , 125mM NaCl], vial of 100mM DTT.

DNA-dependent RNA polymerase with high specificity for bacteriophage T7 promoter sequences

Synthesizes large quantities of RNA from DNA inserted into a transcription vector downstream from a T7 promoter. A T7 RNA Polymerase technical bulletin is available.

  • Application: Synthesis of labeled and unlabeled RNA transcripts.
  • Source: Purified from E. coli expressing the T7 RNA Polymerase gene on a plasmid
  • Performance and Quality Testing: 3' and 5' exodeoxyribonuclease, ribonuclease, and DNA nicking assays; performance in a transcription reaction
  • Unit Definition: One unit hydrolyzes 1nMol of ribonucleotide into acid-precipitable material in 1 hr at 37°C using a T7 transcription vector as template
  • Unit Reaction Conditions: 40mM Tris-HCl (pH 8.0), 25mM NaCl, 8mM MgCl2, 2mM spermidine-(HCl)3, 5mM DTT, 0.4mM ATP, 0.4mM CTP, 0.4mM GTP, 0.4mM UTP, 1μCi [3H]GTP, 1μg SphI-cut pT7L13, and enzyme in 50μL for 10 min. at 37°C

Cell Free Protein Expression for Functional Analysis, Cell Free Protein Expression for Structural Analysis, Cell-Free (in vitro) Expression, Gene Expression Analysis and Genotyping, In Vitro Transcription, Protein Expression, Proteins, Expression, Isolation and Analysis

Informations sur la commande

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Spécification

Content And Storage T7 RNA Polymerase is supplied with a vial of 5X reaction buffer [200 mM Tris-HCl (pH 8.0), 40 mM MgCl2 , 10 mM spermidine-(HCl)3 , 125 mM NaCl], vial of 100 mM DTT. Store at -20°C.
Shipping Condition Wet Ice
Polymerase T7 RNA Polymerase
Promoter T7
Quantity 2500 U

For Research Use Only. Not for use in diagnostic procedures.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.