missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ INVαF' Chemically Competent E. coli
Description
The TA Cloning Kit uses the pCR2.1 cloning vector and ExpressLink T4 DNA Ligase to generate a ligation product in a 15-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.
- Fast and convenient: 15-minute, room-temperature ligation
- Efficient: blue/white screening and >80% clones with correct insert
- Flexible: choice of kanamycin or ampicillin resistance for flexible antibiotic selection
- Hassle-free: eliminates any enzymatic modifications of PCR product
- Streamlined: does not require use of PCR primers that contain restriction sites
- 3'-T overhangs for direct ligation of Taq-amplified PCR products
- T7 promoter for in vitro RNA transcription and sequencing
- Versatile polylinker with flanking EcoR I sites for easy excision of inserts
- M13 forward and reverse primer sites for sequencing
- Taq polymerase has non-template-dependent activity that adds single deoxyadenosine (A) to the 3' ends of PCR products
- Linearized vector supplied in has single 3' deoxythymidine (T) residues
- Allows PCR inserts to ligate efficiently with vector
Cloning, PCR Cloning
Specifications
Specifications
| Bacterial or Yeast Strain | INVαF ́ |
| Cell Type | Chemically Competent E. coli |
| Cloning Method | TA Cloning |
| Content And Storage | TA Cloning™ kits contain linearized pCR™2.1 vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. The competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid. Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
| Format | Kit |
| For Use With (Application) | PCR Cloning |
| No. of Reactions | 40 Reactions |
| Product Line | One Shot |
| Product Type | Cloning Kit |
| Promoter | T7 |
| Show More |
Safety and Handling
Product Identifier
- TA Cloning Kit, with pCR 2.1 Vector and One Shot INV
- Warning
- Reproductive toxicity Category 2
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H361-Suspected of damaging fertility or the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: 2-Mercaptoethanol, Hydrogen chloride
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?