missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10 Chemically Competent E. coli

Product Code. 10564993
Change view
Click to view available options
No. of Reactions:
20 Reactions
40 Reactions
Vector:
pCR2.1
pCRII
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Vector unitSize
10564993 40 Reactions pCR2.1 Each
10093092 20 Reactions pCRII Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 10564993 Supplier Invitrogen™ Supplier No. K204040

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Provides a quick, one-step cloning strategy for directly inserting a Taq-amplified PCR product into a plasmid vector. Invitrogen™ TA Cloning™ Kit, with pCR™2.1 Vector and One Shot™ TOP10 Chemically Competent E. coli is available in 20- and 40-reaction kit sizes.

The TA Cloning™ Kit uses the pCR™2.1 cloning vector and ExpressLink™ T4 DNA Ligase to generate a ligation product in a fifteen-minute, room-temperature ligation step. Reactions typically yield >80% recombinants containing inserts.

  • Fast & Convenient—15-minute, room-temperature ligation
  • Efficient—Blue/white screening and >80% clones with correct insert
  • Flexible—Choice of kanamycin or ampicillin resistance for flexible antibiotic selection
  • Hassle-free—Eliminates any enzymatic modifications of the PCR product
  • Streamlined—Does not require the use of PCR primers that contain restriction sites

The pCR™2.1 vector provides:

  • 3′-T overhangs for direct ligation of Taq-amplified PCR products
  • T7 promoter for in vitro RNA transcription and sequencing
  • Versatile polylinker with flanking EcoR I sites for easy excision of inserts
  • M13 forward and reverse primer sites for sequencing

How TA Cloning™ Works
Taq polymerase has a non-template-dependent activity that adds a single deoxyadenosine (A) to the 3′ ends of PCR products. The linearized vector supplied in this kit has single 3′ deoxythymidine (T) residues. This allows PCR inserts to ligate efficiently with the vector.

Other Kit Configurations
TA Cloning™ Kits are also offered without competent cells (K2020-20, K2020-40), with One Shot™ INVF Chemically Competent E. coli (K2000-01, K2000-40) or with One Shot™ TOP10F Chemically Competent E. coli (K2030-01, K2030-40).

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain TOP10
Cell Type Chemically Competent E. coli
Cloning Method TA Cloning
Content And Storage TA Cloning™ kits contain linearized pCR™2.1 vector, ExpressLink™ T4 DNA ligase, 5X ExpressLink™ T4 DNA ligation buffer, dNTPs, 10X PCR buffer, sterile water, and controls. The competent cell kits contain One Shot™ chemically competent E. coli, S.O.C. medium, and a supercoiled control plasmid.

Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
Format Kit
For Use With (Application) PCR Cloning
No. of Reactions 40 Reactions
Product Line One Shot
Product Type Cloning Kit
Promoter T7
Quantity 40 reactions
Vector pCR2.1
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • TA Cloning Kit, with pCR 2.1 Vector and One Shot TOP10 Chemically Competent E. coli
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-Mercaptoethanol, Hydrogen chloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.