missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOPO™ TA Cloning™ Kit, with One Shot™ MAX Efficiency™ DH5α-T1R E. coli

Product Code. 10624043
Click to view available options
Quantity:
25 Reactions
50 Reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10624043

Brand: Invitrogen™ K452001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Topoisomerase I-activated pCR™2.1-TOPO™ vector; PCR buffer; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Control PCR primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid

Designed for cloning PCR products directly from a PCR reaction in just 5 minutes

They use a pCR™-TOPO Vector with covalently bound topoisomerase I for fast cloning and recombinants.

  • 3'-T overhangs for direct ligation of Taq-amplified PCR products
  • Choice of T7 (pCR2.1-TOPO) or T7 and SP6 (pCRII-TOPO) promoters for in vitro RNA transcription and sequencing. Each vector also contains M13 forward and reverse primer sites for sequencing.
  • EcoR I sites flanking the PCR product insertion site for easy excision of inserts
  • Kanamycin and ampicillin resistance genes for your choice of selection in E. coli
  • Easy blue/white colony screening for selection of recombinants

Cloning, PCR Cloning

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain DH5α
Cloning Method TOPO™-TA
Content And Storage Box 1:
• Topoisomerase I-activated pCR™2.1-TOPO™ vector
• PCR buffer
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Control PCR primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid

Store at -68 to -85°C.
Format Kit
Product Line One Shot™
Product Type Cloning Kit
Promoter T7, SP6
Quantity 25 Reactions
Vector TOPO-TA Cloning Vectors

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.