missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Dithiothreitol

Product Code. 11568736
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11568736 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11568736 Supplier Invitrogen™ Supplier No. 15508013

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

UltraPure Dithiothreitol (DTT, Cleland's reagent), which reduces disulfides to their corresponding thiols, is used at low concentrations to stabilize enzymes containing free sulfhydryl groups.

UltraPure Dithiothreitol (DTT, Cleland's reagent), which reduces disulfides to their corresponding thiols, is used at low concentrations to stabilize enzymes containing free sulfhydryl groups. Higher concentrations of DTT are used to cleave disulfide linkages in polypeptides and to facilitate protein denaturation by detergents or chaotropic agents.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5° to -30°C).
For Use With (Application) Protein Electrophoresis & Western Blotting
Product Line UltraPure
Quantity 5 g
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.