missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Guanidine Hydrochloride

Product Code. 11528736
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11528736

Brand: Invitrogen™ 15502016

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

UltraPure Guanidine Hydrochloride is a strong protein denaturant. In a highly concentrated (5 to 6M) guanidine hydrochloride solution, many proteins unfold and separate into their constituent polypeptide chains.

UltraPure Guanidine Hydrochloride is a strong protein denaturant. In a highly concentrated (5 to 6M) guanidine hydrochloride solution, many proteins unfold and separate into their constituent polypeptide chains. Guanidine hydrochloride is widely used to purify nucleic acids from cell extracts and is useful as an RNase inhibitor during the isolation of mRNA from mammalian cells.

Order Info

Approved for shipment at Room Temperature or on Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) RNA Extraction
Packaging Type Bottle
Product Type Guanidine Hydrochloride
Product Line UltraPure
Purity DNase-Free, RNase-Free
Quantity 500 g
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.