missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Phenol

Product Code. 11578736
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11578736 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11578736 Supplier Invitrogen™ Supplier No. 15509037

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

UltraPure Phenol is used in the purification of nucleic acids. In a mixture of phenol and buffered aqueous solution, proteins are denatured and collect at the interphase while most nucleic acids remain in the aqueous phase.

UltraPure Phenol is used in the purification of nucleic acids. In a mixture of phenol and buffered aqueous solution, proteins are denatured and collect at the interphase while most nucleic acids remain in the aqueous phase. UltraPure Phenol contains no preservatives. It is redistilled and packaged under a dry inert gas in shatter-resistant, plastic-coated amber bottles.

Performance and Quality Testing
Color of water-saturated samples is evaluated after incubation at 2–8°C

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenols
Content And Storage Store in refrigerator (2–8°C).
For Use With (Application) RNA/DNA Extraction
Product Line UltraPure
Purity Molecular Biology Grade
Quantity 500 g
Shipping Condition Room Temperature
Form Powder

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.