missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ SDS Solution, 10%
GREENER_CHOICE

Product Code. 10154042
Click to view available options
Quantity:
1 L
4 x 100 mL
Unit Size:
1 L
400mL
This item is not returnable. View return policy

Product Code. 10154042

Brand: Invitrogen™ 24730020

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Filtered and ready to use

UltraPure 10% SDS Solution is a premixed aqueous solution containing 10% sodium dodecyl sulfate (SDS). SDS is a protein denaturant used in polyacrylamide gel electrophoresis for molecular weight determinations.

  • Used to dissociate nucleic acid-protein complexes in DNA extraction protocols, to disrupt cell membranes, and to prepare prehybridization and hybridization solutions
  • Sustainable packaging

DNA & RNA Purification & Analysis, DNA Extraction, Gene Expression Analysis & Genotyping, General gDNA Purification Reagents and Accessories, Genomic DNA Purification, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting, Southern Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 bottle containing 1 L of Ultrapure SDS at 10% concentration (w/v). Store at room temperature.
Concentration 0.1
Product Type SDS Solution
Form Liquid
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Product Line UltraPure™
Quantity 1 L
Reagent Type SDS
Shipping Condition Room Temperature
compliance-icons
Product Identifier
  • UltraPure SDS Solution, 10%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : Sodium lauryl sulphate

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.