missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ Sodium Dodecyl Sulfate (SDS)
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
- Used in determination of protein molecular weight by polyacrylamide gel electrophoresis
- Used to disrupt cell walls and dissociate nucleic acid-protein complexes in DNA and RNA extraction protocols
DNA & RNA Purification & Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting
Order Info
Shipping Condition: Approved for shipment at room temperature or on wet ice
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Product Type | Sodium Dodecyl Sulfate |
| Form | Powder |
| For Use With (Application) | Nucleic Acid Extraction |
| Product Line | UltraPure™ |
| Quantity | 500 g |
| Reagent Type | SDS |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction