missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ SSC, 20X

Product Code. 11598976
Change view
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11598976 4 L 4 L
10515203 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11598976 Supplier Invitrogen™ Supplier No. 15557036
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in nucleic acid hybridizations and blot transfer applications

UltraPure 20X SSC is a solution formulated and used in concentrations ranging from 0.2X to 20X, depending on the application.

  • Supplied in 1 L plastic bottles/4 L/10 L stackable Cubitainer™ Box
  • Sterile-filtered UltraPure 20X SSC contains 3.0 M NaCl and 0.3 M sodium citrate, at pH 7.0
  • Performance and Quality Testing: No DNase or RNase activity detected

DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Concentration 20 X
Form Liquid
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
Quantity 4 L
Shipping Condition Room Temperature
Product Line UltraPure
Product Type SSC

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.