missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ SSPE, 20X

Click to view available options
Quantity:
1 L
10 L
Unit Size:
1 L
10 L
Description
- UltraPure 20X SSPE is prepared buffer concentrate
- Supplied in 1 L plastic bottles/4 L/10 L stackable Cubitainer™, Box
- Sterile-filtered UltraPure 20X SSPE contains 3.0 M NaCl, 0.2M NaH2PO4, and 0.02M EDTA at pH 7.4
- Performance and Quality Testing: No DNase or RNase activity detected. Sustainable packaging
DNA & RNA Purification & Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Concentration | 20 X |
| Content And Storage | Store at room temperature. |
| For Use With (Application) | Nucleic Acid Gel Electrophoresis, Blotting |
| Product Line | UltraPure |
| Product Type | SSPE |
| Quantity | 1 L |
| Shipping Condition | Room Temperature |
| Form | Liquid |
Safety and Handling
Product Identifier
- UltraPure SSPE, 20X
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contains : Sodium hydroxide
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction