missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ Low Melting Point Agarose
GREENER_CHOICE

Artikelnummer. 10032744
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
100 g
missing translation for 'unitSize'
100g
50g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 10032744

missing translation for 'mfr': Invitrogen™ 16520100

Please to purchase this item. Need a web account? Register with us today!

Edge
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Easy-pour spout reduces spills and contamination

UltraPureLow Melting Point Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPureLow Melting Point Agarose is ideal for resolving DNA and RNA fragments larger than 1,000bp and for the recovery of nucleic acid fragments after electrophoresis, as it melts at 65.5°C (below the melting point of most nucleic acids). The solution remains fluid at 37°C and will set rapidly at temperatures below 25°C

  • Ideal for analysis and recovery of DNA and RNA for routine applications
  • Strong gel structure allows for better handling and less breakage
  • Ideal for variety of applications such as in-gel manipulation, tissue and cell culture, and viral plaque assays
  • Can also be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)
  • Improved packaging: Environmentally friendly, sustainable packaging uses 75% less plastic than the original bottles. This means less energy and raw material used in manufacture, and less waste in Landfills
  • Performance and quality testing: Performance of UltraPure Agarose is evaluated to satisfy specifications in appearance, moisture, gel strength, gelling temperature, melting temperature, sulfate content, electroendosmosis, and DNase/RNase assay
  • Separation range: >1,000 bp.

UltraPureLow Melting Point Agarose (Cat. No. 16520-100) is a replacement for the item previously sold under Cat. No. 15517-022.

Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting

Bestellinformation

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage 1 pouch containing 100 g of UltraPure™ Low Melting Point Agarose. Store at room temperature.
Product Type Agarose
Separation Range 100 to >30000 bp
Form Powder
For Use With (Application) Nucleic Acid Gel Electrophoresis, Protein Electrophoresis, Blotting
Gel Compatibility Agarose Gels
Green Features Sustainable packaging
Melting Point Low Melting Point
Product Line UltraPure
Quantity 100 g
Shipping Condition Room Temperature
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt