missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ Urea
Click to view available options
Quantity:
500 g
2 kg
Product Type:
UltraPure Urea
Urea
Unit Size:
2kg
Each
Description
UltraPure™ Urea is an effective protein denaturant. In a highly concentrated (6 to 8 M) urea solution, many proteins unfold and separate into their constituent polypeptide chains. UltraPure™ Urea is also used to denature nucleic acids in sequencing gels.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Storage conditions: 15-30°C |
| Product Type | Urea |
| Product Line | UltraPure |
| Quantity | 2 kg |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
| Form | Powder |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction