missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Uracil DNA Glycosylase

Product Code. 10748653
Change view
Click to view available options
Quantity:
100 units
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10748653 100 units Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10748653 Supplier Invitrogen™ Supplier No. 18054015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Removes uracil residues from sugar moiety of single- and double-stranded DNA without destroying phosphodiester backbone

Uracil DNA Glycosylase (uracil-N-glycosylase) removes uracil residues from the sugar moiety of single- and double-stranded DNA without destroying the phosphodiester backbone, preventing its use as a hybridization target or as a template for DNA polymerases.

Also known as uracil-N-glycosylase.

  • Does not remove uracil from RNA
  • Purified from E. coli expressing E. coli ung gene on plasmid
  • One unit catalyzes the release of 1nmol of free uracil from 3H-poly(dU) in 1 hr. at 37°C

Helps to eliminate carryover contamination in PCR, resulting in fewer false positive results for cloning PCR fragments

Recommended Storage

Store at -5deg; to -30°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5° to -30°C).
Form Liquid
PCR Method qPCR
Polymerase DNA Polymerase
Quantity 100 units
Shipping Condition Approved for shipment on Wet or Dry Ice
For Use With (Application) Standard PCR, Real Time PCR (qPCR)
What is UDG?

UDG (uracil-DNA-glycosylase) refers to a superfamily of enzymes comprising six sub-families. Family I UDG enzymes are called UNG, after the uracil-N-glycosylase gene. The terms UDG and UNG are commonly used interchangeably because they perform the same function in qPCR, namely, to remove Uracil from dU-containing DNA to prevent carryover contamination from previous PCRs. See the following link: https://www.thermofisher.com/uk/en/home/life-science/pcr/real-time-pcr/real-time-pcr-learning-center/real-time-pcr-basics/what-is-ung-udg.html

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.