missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViraPower™ Adenoviral Gateway™ Expression Kit

Product Code. 10296422
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10296422 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10296422 Supplier Invitrogen™ Supplier No. K493000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Simplifies cloning for greater efficiency in generating high-titer, recombinant adenovirus

  • Facilitates highly efficient, in vitro or in vivo delivery of target gene to dividing and nondividing mammalian cells using replication-incompetent adenovirus
  • pAd/CMV/V5-DEST Gateway-adapted adenoviral vector permits high-level protein expression from human cytomegalovirus (CMV) immediate-early enhancer/promoter as well as 293A cell line
  • Produce high titer adenoviral stocks up to 1 x 1011 pfu/mL in concentrated preparations
  • Bypasses inefficient homologous recombination in human or bacterial cells
  • Amenable for high-throughput applications or library transfer applications
  • Replication-incompetent virus enhances biosafety of the system and its use as gene delivery vehicle

Antibody and Cell Culture Production, Bioproduction, Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery, Viral Vaccine Production

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Adenoviral
Promoter CMV
Product Type Adenoviral Expression Kit
Content And Storage pAd⁄CMV⁄V5-DEST Gateway™ Vector: store at -20°C;.
293A Cells: store in liquid nitrogen.
Guaranteed stable for 6 months when properly stored.
Protein Tag V5 Epitope Tag
Cloning Method Gateway
Quantity 1 kit
Vector pAd
Product Line Gateway, ViraPower
For Use With (Application) Viral Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.