missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ViraPower™ Lentiviral Directional TOPO™ Expression Kit

Product Code. 17195989
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17195989 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17195989 Supplier Invitrogen™ Supplier No. K495000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Includes

pLenti6/V5 Directional TOPO Cloning Kit: pLenti6/V5 Directional TOPO Reagents, Stbl3 Chemically Competent E. coli, ViraPower Bsd Lentiviral Support Kit: ViraPower Packaging Mix, Lipofectamine 2000, Blasticidin, 293FT Cell Line

Facilitates quick PCR-based cloning and lentiviral-based expression of target gene in dividing and nondividing mammalian cells

  • Includes all components needed to generate lentivirus, including vector kit, 293FT cell line, and support kit
  • PLenti6/V5-D-TOPO vector has the CMV promoter for driving constitutive expression of the target gene and blasticidin selection marker for stable selection in mammalian cells
  • Constitutive high expression with CMV promoter
  • Blasticidin selection marker for stable selection
  • C terminal V5 tag for quick detection

Constitutive Expression, Mammalian Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Viral Gene Delivery

TRUSTED_SUSTAINABILITY

Specifications

Constitutive or Inducible System Constitutive
Delivery Type Lentiviral
Promoter CMV
Product Type Lentiviral Expression Kit
Selection Agent (Eukaryotic) Blasticidin
Content And Storage pLenti6⁄V5™ Directional TOPO™ Cloning Kit:
• pLenti6⁄V5™ Directional TOPO™ Reagents: -20°C
• One Shot™ Stbl3™ Chemically Competent E. coli:-80°C

ViraPower™ Bsd Lentiviral Support Kit:
• ViraPower™ Packaging Mix: -20°C
• Lipofectamine™ 2000: 4°C (DO NOT freeze)
• Blasticidin: -20°C
• 293FT Cell Line: liquid nitrogen
Protein Tag V5 Epitope Tag
Cloning Method Directional TOPO
Quantity 1 kit
Vector Directional TOPO Vectors
Product Line One Shot
For Use With (Application) Viral Expression
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.