missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast Deletion Mat, Complete Set

Product Code. 10175114
Change view
Click to view available options
Quantity:
53 Plates
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10175114 53 Plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10175114 Supplier Invitrogen™ Supplier No. 95401.H2

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Yeast deletion library created by a collaboration of eight North American and eight European laboratories composing the Saccharomyces Genome Deletion Project

Each yeast deletion clone is supplied as a stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418).

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Yeast Deletion Mat
Libraries Yeast deletion library created by a collaboration of 8 North American and 8 European laboratories composing the Saccharomyces Genome Deletion Project
Culture Type Stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418)
Format Complete Set
Quantity 53 Plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.