missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast tRNA

Product Code. 11508736
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11508736 25 mg 25mg
11518736 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11508736 Supplier Invitrogen™ Supplier No. 15401011
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Supplied in lyophilized form

Yeast tRNA is suitable for use as a carrier in nucleic acid purification and precipitation procedures.

Array CGH, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Microarray Analysis

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Yeast tRNA
Content And Storage Store in freezer (-5 to -30°C).
Reagent Type Yeast tRNA
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
For Use With (Application) Carrier for Nucleic Acid Precipitation
Quantity 25 mg
Form Lyophilized Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.