Learn More
Invitrogen™ Zero Blunt™ TOPO™ PCR Cloning Kit for Sequencing, with One Shot™ TOP10 Chemically Competent E. coli
Description
Includes
Topoisomerase I-activated pCR™4Blunt-TOPO™ vector; Salt solution; dNTPs; Control template; T3, T7, M13 forward, and M13 reverse primers; Control PCR primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid
Each kit uses the pCR™4Blunt-TOPO vector with specially designed sequencing primer sites that return more insert sequence and less vector sequence from each reaction. Kits includes everything necessary to clone and select recombinant vectors containing your PCR fragment of choice.
pCR4Blunt-TOPO vector: optimized for sequencing
We have removed much of the multiple cloning site from the pCR4Blunt-TOPO vector to shorten the distance between sequencing primer sites and the insert site to as little as 33 bp. This means sequencing reactions give less vector sequence and more insert sequence. The pCR4Blunt TOPO vector has sites for four common sequencing primers: M13 forward, M13 reverse, T7, and T3. The kits include an aliquot of each.
pCR4Blunt-TOPO clone selection and manipulation
The pCR4Blunt-TOPO vector contains both ampicillin and kanamycin resistance markers and a LacZα-ccdB gene fusion for positive selection and blue/white screening. The vector's minimized multiple cloning site still includes flanking EcoRI sites for simplified excision of cloned PCR products and a unique Sse8387I site for generation of nested deletions prior to sequencing. T7 and T3 promoters are also present for in vitro transcription.
Simplified, TOPO-based cloning
Using TOPO cloning technology, there is no need for PCR primers containing specific sequences, post-PCR procedures, vector preparation, or other time-intensive DNA manipulation steps. Just add your PCR reaction straight to the provided topoisomerase-charged vector, incubate 5 minutes, and transform the provided E. coli competent cells.
Efficient cloning
With up to 95% of clones carrying the desired insert, you can screen fewer clones, which saves you time and money. The pCR4Blunt-TOPO vector used in this kit comes with no overhangs for efficient ligation of PCR products created by proofreading, thermostable polymerases that leave blunt-ended PCR products.
The standard in cloning
When it comes to cloning, TOPO cloning technology has been a reliable partner for thousands of scientists for over ten years. Fast, simple-to-use, and efficient, TOPO cloning has been applied to many different vectors for a wide array of applications.
Zero Blunt TOPO PCR Cloning Kit for Sequencing: kit formats
The Zero Blunt; TOPO PCR Cloning Kit for Sequencing can be purchased with a variety of competent cells that deliver different advantages depending on what your needs are:
- General cloning: TOP10 cells (Cat. Nos. K2875-J10, K2875-20, K2875-40)
- High-efficiency cloning: TOP10 Electrocomp™ Cells (Cat. Nos. K2880-20, K2880-40)
- General cloning, bacteriophage T1 resistance: DH5α-T1R (Cat. No. K2895-20)
- Fast growth: Mach1™-T1R Chemically Competent E. Coli (Cat. No. K2835-20)
Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Cloning, Methylation Analysis, PCR Cloning, RNAi, Epigenetics and Non-Coding RNA Research, Sequencing Bisulfite-Treated DNA
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Bacterial or Yeast Strain | TOP10 |
| Cell Type | Chemically Competent |
| Cloning Method | Blunt TOPO™ |
| Content And Storage | Linearized and topoisomerase I-activated pCR™4-Blunt TOPO™ Vector, Salt Solution, dNTPs, Control Template and Primers, M13 Forward and Reverse Primers, One Shot™ Chemically Competent TOP10 E. coli, S.O.C. Medium, and a supercoiled control plasmid. Store One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored. |
| Format | Kit |
| For Use With (Application) | Chromatin Biology |
| Product Line | One Shot |
| Product Type | PCR Cloning Kit |
| Promoter | T7, T3 |
| Quantity | 10 Reactions |
| Afficher plus |
Frequently Asked Questions (FAQs)
We suggest starting with a molar ratio of 1:1 (insert:vector), with a range of 0.5:1 to 2:1. The quantity used in a TOPO cloning reaction is typically 5-10 ng of a 2 kb PCR product.
Equation:
length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 (insert:vector ratio)
The optimal ratio is 1:1 insert to vector. Optimization can be done using a ratio of 0.5-2 molecules of insert for every molecule of the vector.
Equation:
length of insert (bp)/length of vector (bp) x ng of vector = ng of insert needed for 1:1 insert:vector ratio
Using the vector only for transformation is not a recommended negative control. The process of TOPO-adaptation is not a 100% process, therefore, there will be vector only present in your mix, and colonies will be obtained.
Phosphorylated products can be TA cloned but not TOPO cloned. This is because the necessary phosphate group is contained within the topoisomerase-DNA intermediate complex of the vector. TOPO vectors have a 3' phosphate to which topoisomerase is covalently bound and a 5' phosphate. Non-TOPO linear vectors (TA and Blunt) have a 3' OH and a 5' phosphate. Phosphorylated products should be phosphatased (CIP) before TOPO cloning.
You may be cloning in an artifact. TA and TOPO Cloning are very efficient for small fragments (< 100 bp) present in certain PCR reactions. Gel-purify your PCR product using either a silica-based DNA purification system or electroelution. Be sure that all solutions are free of nucleases (avoid communal ethidium bromide baths, for example.)
For Research Use Only. Not for use in diagnostic procedures.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.