missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zero Blunt™ TOPO PCR Cloning Kit for Subcloning

Product Code. 10434012
Change view
Click to view available options
Quantity:
25 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10434012 25 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10434012 Supplier Invitrogen™ Supplier No. K283020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Linearized and topoisomerase I-activated pCR™Blunt II-TOPO™ vector; Salt solution; dNTPs; Control template; M13 forward and reverse primers; Sterile water; One Shot™ Chemically Competent or Electrocomp™ E. coli; S.O.C. medium; Supercoiled pUC19 control plasmid

Combines the unique Zero Background™ technology with the pCR™- Blunt II-TOPO vector to allow easy, high-efficiency cloning of blunt-end PCR products

  • Unique technology to minimize background
  • Get your clone: 95% efficiency
  • Multiple primer sites (T7, T3, M13F, M13R) make sequence analysis or PCR easy and convenient
  • Kanamycin and Zeocin resistance genes for your choice of selection
  • Convenient, validated restriction sites in MCS for further subcloning
  • Includes MachI™ T1R competent cells for fast miniprep and shortens cloning time

Chromatin Biology, Chromatin Immunoprecipitation (ChIP), Cloning, Methylation Analysis, PCR Cloning, RNAi, Epigenetics and Non-Coding RNA Research, Sequencing Bisulfite-Treated DNA

TRUSTED_SUSTAINABILITY

Specifications

Bacterial or Yeast Strain Mach1™
Cloning Method Blunt TOPO™
Content And Storage Box 1:
• Linearized and topoisomerase I-activated pCR™Blunt II-TOPO™ vector
• Salt solution
• dNTPs
• Control template
• M13 forward and reverse primers
• Sterile water

Store at -5 to -30°C.
All reagents are stable for 6 months when properly stored.

Box 2:
• One Shot™ Chemically Competent or Electrocomp™ E. coli
• S.O.C. medium
• Supercoiled pUC19 control plasmid

Store at -68 to -85°C.
Format Kit
For Use With (Application) Chromatin Biology
Product Line One Shot
Product Type PCR Cloning Kit
Quantity 25 Reactions
Vector Blunt DNA Cloning Vectors

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.