missing translation for 'onlineSavingsMsg'
Learn More

Iodic acid, 99.5%, for analysis

Product Code. 10606432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10606432 50 g Glass bottle 50g
10409910 250 g Glass bottle 250g
10590964 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10606432 Supplier Thermo Scientific Acros Supplier No. 206450500
  • ONLINE EXCLUSIVE PRICE
    54.10 €
    52.50 € / 50g
    Save 1.60 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7782-68-5 | ILiO3 | 181.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-68-5
Molecular Formula ILiO3
Molecular Weight (g/mol) 181.84
MDL Number MFCD00011348
InChI Key FZAXZVHFYFGNBX-UHFFFAOYSA-M
Synonym unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
PubChem CID 24345
ChEBI CHEBI:24857
IUPAC Name lithium(1+) iodate
SMILES [Li+].[O-][I](=O)=O

Specifications

Color White to Beige
Melting Point 110.0°C
Density 4.6200g/mL
Heavy Metals (as Pb) 10ppm max.
Packaging Glass bottle
Specific Gravity 4.62
Assay Percent Range 99.5%
Quantity 50 g
Linear Formula HIO3
Physical Form Crystalline Powder
Merck Index 15, 5056
Solubility Information Solubility in water: 2690g/l (20°C). Other solubilities: soluble in nitric acid, soluble in diluted alcohol, insoluble in absolute alcohol, ether and, chloroform
Formula Weight 175.91
Percent Purity 99.5%
Grade Analytical
Chemical Name or Material Iodic acid, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.