missing translation for 'onlineSavingsMsg'
Learn More

Iodic acid, 99.6%, ACS reagent

Product Code. 10751452 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10751452 100 g Glass bottle 100g
10509413 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10751452 Supplier Thermo Scientific Acros Supplier No. 423811000
  • ONLINE EXCLUSIVE PRICE
    90.25 €
    83.40 € / 100g
    Save 6.85 €
    7% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7782-68-5 | ILiO3 | 181.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-68-5
Molecular Formula ILiO3
Molecular Weight (g/mol) 181.84
MDL Number MFCD00011348
InChI Key FZAXZVHFYFGNBX-UHFFFAOYSA-M
Synonym unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min
PubChem CID 24345
ChEBI CHEBI:24857
IUPAC Name lithium(1+) iodate
SMILES [Li+].[O-][I](=O)=O

Specifications

Color White
Melting Point 110.0°C
Density 4.6200g/mL
Heavy Metals (as Pb) 10ppm max.
Packaging Glass bottle
Residue after Ignition 0.02% max.
Specific Gravity 4.62
Assay Percent Range 99.6%
Linear Formula HIO3
Quantity 100 g
Physical Form Crystalline Powder
Merck Index 15, 5056
Solubility Information Solubility in water: 2690g/l (20°C). Other solubilities: soluble in nitric acid, soluble in diluted alcohol, insoluble in absolute alcohol, ether and, chloroform
Formula Weight 175.91
Percent Purity 99.6%
Grade ACS Reagent
Chemical Name or Material Iodic acid, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.