missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodide Iodate concentrate, For 1L standard solution, 0.005 M I2 (0.01N), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 100°C |
| Physical Form | Concentrate |
| Quantity | 6 x 1 Ea. |
| UN Number | NONH for all modes of transport |
| Beilstein | 3587194 |
| Packaging | Ampoule |
| Density | 1.000 g/cm3 |
| Chemical Name or Material | Iodide Iodate concentrate, For 1L standard solution, 0.005 M I2 (0.01N) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction