missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 99.5+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 10500921 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10500921 250 g 250g
10694722 100 g 100g
10694912 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10500921 Supplier Fisher Chemical Supplier No. I/0450/50
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Boiling Point 185°C
Melting Point 113°C
Color Gray
pH 5.1
Physical Form Solid
Quantity 250 g
IUPAC Name molecular iodine
Formula Weight 253.81g/mol
Packaging Amber glass bottle
Vapor Pressure 0.41 hPa 25
Density Vapor Density: 8.8
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Iodine
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P273-Avoid release to the environment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.