missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Iodine Determination Flasks with Conical Ground Joint, without Collar

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 03026052

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10718651

  • 241.68 € / Pack of 10
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

  • With hollow hexagonal stopper NS 29/32
  • With scale (approx. volume /ml)
  • With inscription label
  • Material: DURAN™
Specifications

Specifications

300 mL
10/Pk.
87 mm
Duran Glass
NS 29/32
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers