missing translation for 'onlineSavingsMsg'
Learn More

Iodine Indicator, Technical, Indicator Grade, Fisher Chemical™

Product Code. 10614922 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10614922 250 g 250g
12627915 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10614922 Supplier Fisher Chemical Supplier No. I/0505/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Iodine Indicator, Technical, Indicator Grade

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 3703398

Specifications

Packaging Plastic powder jar
Color White
pH 5 to 7
Quantity 250 g
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.