missing translation for 'onlineSavingsMsg'
Learn More

Iodine lump, ultra dry, 99.999% (metals basis)

Product Code. 15482047
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482047 5 g 5g
15492047 25 g 25g
15402057 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482047 Supplier Thermo Scientific Alfa Aesar Supplier No. 044857.06
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 7553-56-2 | I2 | 253.81 g/mol

Iodine lump is used in OLED materials, pharmaceuticals, and the electronics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
IUPAC Name diiodine
SMILES II

Specifications

Melting Point 183°C to 186°C (sublimation)
Physical Form Solid
Quantity 5 g
Percent Purity 99.999%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.