missing translation for 'onlineSavingsMsg'
Learn More

Iodine monobromide, 98%

Product Code. 10125453 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle in plastic container
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10125453 250 g Glass bottle in plastic container 250g
10522231 50 g Glass bottle in plastic container 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10125453 Supplier Thermo Scientific Acros Supplier No. 208842500
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 7789-33-5 | BrI | 206.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-33-5
Molecular Formula BrI
Molecular Weight (g/mol) 206.81
MDL Number MFCD00011353
InChI Key CBEQRNSPHCCXSH-UHFFFAOYSA-N
Synonym iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
PubChem CID 82238
IUPAC Name iodobromane
SMILES BrI

Specifications

Boiling Point 116.0°C
Melting Point 42.0°C to 50.0°C
Color Black-Brown to Gray
Physical Form Crystalline Mass, Crystals or Chunks
Quantity 250 g
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula IBr
Fieser 01,501; 02,224
Merck Index 15, 5059
Solubility Information Solubility in water: soluble. Other solubilities: solubel in alcohol,ether,carbon disulfide and,glacial acetic acid
Formula Weight 206.81
Percent Purity 98%
Packaging Glass bottle in plastic container
Specific Gravity 4.41
Density 4.4100g/mL
Chemical Name or Material Iodine monobromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodine monobromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.