missing translation for 'onlineSavingsMsg'
Learn More

Iodine Monochloride, ACS Reagent, 1.10±0.1 I/Cl Ratio Basis, Honeywell™ Fluka™

Product Code. 15665420 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15665420 25 g Glass Bottle 25g
15655420 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15665420 Supplier Honeywell-Fluka Supplier No. 40299025G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Boiling Point >97°C
Melting Point 14°C to 27°C
Color Brown
Quantity 25 g
Linear Formula ICl
UN Number UN1792
Formula Weight 162.36g/mol
Percent Purity 1.10 ± 0.1% (I/Cl ratio basis)
Odor Stinging
Packaging Glass Bottle
Appearance Liquid or Solid
Density 3.24g/mL at 25°C
Chemical Name or Material Iodine monochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.