missing translation for 'onlineSavingsMsg'
Learn More

Iodine monochloride, ACS reagent

Product Code. 10108040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10108040 100 g Glass bottle 100g
10371781 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10108040 Supplier Thermo Scientific Acros Supplier No. 412361000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-99-0 | ClI | 162.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Boiling Point 97.0°C
Melting Point 26.0°C
Color Brown-Red
Physical Form Fuming Liquid
Quantity 100 g
Linear Formula ICl
Fieser 01,502; 08,261; 10,212; 11,268
Merck Index 15, 5060
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes, soluble in ethanol, ether and cs2
Formula Weight 162.36
Percent Purity 1.0 to 1.2 (I/Cl ratio of the WIJS solution prepared from the reagent)
Grade ACS Reagent
Packaging Glass bottle
Specific Gravity 3.2
Density 3.2000g/mL
Chemical Name or Material Iodine monochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.