missing translation for 'onlineSavingsMsg'
Learn More

Iodine Monochloride, Reagent grade, ≥95%, Honeywell™

Product Code. 15661830 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass Bottle
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15661830 5 g Glass Bottle 5g
15641830 100 g Glass Bottle 100g
15651830 250 g Glass Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15661830 Supplier Honeywell Research Chemicals Supplier No. 2082215G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Boiling Point >97°C
Melting Point 27°C
Color Brown to Red
Physical Form Liquid or Solid
Quantity 5 g
Linear Formula ICl
UN Number UN1792
Formula Weight 162.36g/mol
Percent Purity ≥95%
Grade Reagent
Packaging Glass Bottle
Density 3.24g/mL at 25°C
Chemical Name or Material Iodine Monochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.