missing translation for 'onlineSavingsMsg'
Learn More

Iodine, puriss., Solstice
BIOPHARMA

Product Code. 15690650 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
6 x 100 g
6 x 1 kg
Unit Size:
100g
1kg
25g
500g
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15690650 1 kg 1kg
15600660 25 g 25g
15680650 100 g 100g
15610660 500 g 500g
15620660 6 x 100 g Pack of 6
15630660 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15690650 Supplier Honeywell-Fluka Supplier No. 030021KG
  • ONLINE EXCLUSIVE PRICE
    415.00 €
    389.76 € / 1kg
    Save 25.24 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.8-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Quantity 1 kg
UN Number 3495
Beilstein 3587194
IUPAC Name diiodine
Formula Weight 253.81g/mol
Percent Purity 99.8 to 100.5%
Grade puriss.
Vapor Pressure 0.31mmHg (25°C), 1mmHg (38.7°C)
Density Vapor Density: 9 (vs air)
Chemical Name or Material Iodine, Meets analytical specification of Ph. Eur.BP, USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.