missing translation for 'onlineSavingsMsg'
Learn More

Iodine, Reagent Grade, beads, 1-3 mm, 99.7% trace metals basis, Honeywell™

Product Code. 15632810 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15632810

Brand: Honeywell Research Chemicals 2664261KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, beads, 1-3 mm, 99.7% trace metals basis

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name molecular iodine
SMILES II

Specifications

Boiling Point 184°C (literature)
Melting Point 113°C (literature)
CAS Max % 100.0000%
Physical Form Beads
Quantity 1 kg
Linear Formula NaH2PO2 · xH2O
UN Number UN3495
Beilstein 3587194
Formula Weight 253.81
Percent Purity ≥99.7%
Grade Reagent
Packaging Glass Bottle
Density 4.930 g/cm3
Chemical Name or Material Iodine, Reagent Grade, beads, 1-3 mm, 99.7% trace metals basis
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.