missing translation for 'onlineSavingsMsg'
Learn More

Iodine Solution, SPEX CertiPrep™

Product Code. 16311136 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 gal.
Packaging:
Amber Glass Bottle
Unit Size:
4.54 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
16311136 1 gal. Amber Glass Bottle 4.54 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16311136 Supplier SPEX Certiprep Supplier No. 183134

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Guaranteed clean and stable, the matrix is prepared using double-deionized, ASTM Type I Water. SPEX CertiPrep™ Iodine solutions are prepared from ACS Grade potassium iodide and crystalline elemental iodine.

  • Ideal for ASTM D1510 applications
  • Packaged in an amber glass bottle

Specifications

CAS 7681-11-0, 7732-18-5
Physical Form Liquid
Quantity 1 gal.
Assay Percent Range Iodine: 99.8%, Potassium Iodide: 99%
Molecular Formula IK
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
IUPAC Name potassium;iodide
Molecular Weight (g/mol) 166.003
PubChem CID 4875
ChEBI CHEBI:8346
Formula Weight Iodine: 126.90447 u, Potassium Iodide: 166.0028 g/mol
Concentration or Composition (by Analyte or Components) 0.0473 N
Packaging Amber Glass Bottle
Chemical Name or Material H2O, KI
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.