missing translation for 'onlineSavingsMsg'
Learn More

Iodine Solution, Wij'S, Pure, Indicator Grade, Fisher Chemical™

Product Code. 10548230 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10548230 1 L 1 L
10476870 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10548230 Supplier Fisher Chemical Supplier No. J/4500/PB15
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

2.5L, 118 deg.C, Amber, 17 deg.C, 11354, Plastic Coated Glass bottle, Liquid, 2.1, 11mmHg at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 11354

Specifications

Boiling Point 118°C
Melting Point 17°C
Color Amber
pH 2
Physical Form Liquid
Quantity 1 L
Packaging Plastic Coated Glass bottle
Vapor Pressure 11mmHg at 20°C
Density Vapor Density: 2.1
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Iodine solution - Wij``s
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Acetic acid, Iodinemonochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.