missing translation for 'onlineSavingsMsg'
Learn More

Iodine, TRC

Product Code. 30405199
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405199 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405199 Supplier Toronto Research Chemicals Supplier No. TRCI6667505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iodine
CAS 7553-56-2
Quantity 5 g
Molecular Formula I2
Synonym Iodine, Actomar, DentaPure DP 90, Diatomic iodine, Diiodine, Eranol, Iodel FD, Iodine (127I2), Iodine Colloidal, Iodine Crystals, Iodine Molecule (I2), Iodine Sublimed, Iosan Superdip, Jodosan, Molecular Iodine, NSC 42355, Tegodyne, eXIA 160
InChI Formula InChI=1S/I2/c1-2
SMILES II
IUPAC Name molecular iodine
Molecular Weight (g/mol) 253.81
Formula Weight 253.8089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.