missing translation for 'onlineSavingsMsg'
Learn More

Iodine trichloride, 97%

Product Code. 15482418
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482418 5 g 5g
15492418 25 g 25g
15402428 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482418 Supplier Thermo Scientific Alfa Aesar Supplier No. H56005.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 865-44-1 | Cl3I | 233.254 g/mol

Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine trichloride can be used to introduce iodine and chlorine in organic synthesis. It is also used as topical antiseptic and as laboratory reagent.

Solubility
Soluble with decomposition.

Notes
Light Sensitive. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-44-1
Molecular Formula Cl3I
Molecular Weight (g/mol) 233.254
MDL Number MFCD00036291
InChI Key PAWIVBWALDNUJP-UHFFFAOYSA-N
Synonym iodine trichloride, iodine chloride icl3, unii-1e5kq66trq, 1e5kq66trq, jodtrichlorid, trichloro-, e3-iodane, trichloro-$l^ 3-iodane, ksc450m5f, iodine trichloride, saj first grade
PubChem CID 70076
IUPAC Name trichloro-$l^{3}-iodane
SMILES ClI(Cl)Cl

Specifications

Quantity 5 g
Linear Formula ICl3
UN Number UN3085
Merck Index 14,5020
Solubility Information Soluble with decomposition.
Formula Weight 233.26
Percent Purity 97%
Sensitivity Light sensitive
Chemical Name or Material Iodine trichloride
compliance-icons compliance-icons
Product Identifier
  • Iodine trichloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Iodine chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.