missing translation for 'onlineSavingsMsg'
Learn More

Iodine(V) oxide, 98%

Product Code. 11365019
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365019 25 g 25g
11375019 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365019 Supplier Thermo Scientific Alfa Aesar Supplier No. 087682.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An oxidizing agent to remove CO from gas | CAS: 12029-98-0 | I2O5 | 333.804 g/mol

Iodine (V) oxide is used as an oxidizing agent to remove carbon monoxide from gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12029-98-0
Molecular Formula I2O5
Molecular Weight (g/mol) 333.804
MDL Number MFCD00011356
InChI Key BIZCJSDBWZTASZ-UHFFFAOYSA-N
Synonym iodine pentoxide, iodopentoxide, iodic anhydride, iodine v oxide, diiodine pentaoxide, unii-upu35c0q0n, upu35c0q0n, iodine oxide, jodpentoxyd, iodine pentaoxide
PubChem CID 159402
ChEBI CHEBI:29914
IUPAC Name iodyl iodate
SMILES O=I(=O)OI(=O)=O

Specifications

Melting Point 275°C to 350°C (decomposition)
Color White to Cream
pH 1.1
Physical Form Crystalline
Quantity 25 g
UN Number UN3085
Merck Index 14,5019
Solubility Information Soluble in water and nitric acid; Insoluble in absolute alcohol,chloroform,ethanol,ether and carbon disulfide.
Formula Weight 333.81
Percent Purity 98%
Sensitivity Hygroscopic; Light sensitive
Density 5.08 g/mL
Chemical Name or Material Iodine (V) oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iodine(V) oxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Iodine oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.