missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Ultrapure, proteomic-grade iodoacetamide is used for efficient alkylation of thiols while minimizing oxidation of the competing thiol pairs in protein sample
Specifications
Specifications
| For Use With (Application) | For efficient alkylation of thiols while minimizing oxidation of competing thiol pairs in Protein samples |
| Quantity | 25000 mg |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 4
- H301-Toxic if swallowed.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- H413-May cause long lasting harmful effects to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?