missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Iodoacetamide Azide

Product Code. 10143454
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10143454 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10143454 Supplier Molecular Probes™ Supplier No. I10188

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

iodoacetamide azide

Conjugates prepared with the thiol-reactive azide, succinimidyl ester can be detected with an alkyne-containing molecule in a click chemistry reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Quantity 1 mg
Product Type Iodoacetamide Azide
Molecular Weight (g/mol) 310.14 Da
Solubility DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C, desiccated and protected from light.
Chemical Reactivity Thiol
Reactive Group Iodoacetamide
Shipping Condition Room Temperature
Reactive Moiety Alkyl Halide, Iodoacetamide
Label or Dye Azide
Label Type Click Chemistry Label
Product Line Molecular Probes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.