missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Iodoacetamido-LC-Phosphonic Acid (6C-CysPAT)

Product Code. 17691556 Shop All Gibco Products
Change view
Click to view available options
Quantity:
10 x 3.5 mg
Unit Size:
35mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17691556 10 x 3.5 mg 35mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17691556 Supplier Thermo Scientific™ Supplier No. A52285
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Iodoacetamido-LC-Phosphonic Acid (6C-CysPAT) is a sulfhydryl-reactive alkylating reagent that contains a phosphonic acid group for enrichment of cysteine-containing peptides for mass spectrometry analysis.

  • A sulfhydryl-reactive alkylating reagent that contains a phosphonic acid group for enrichment of cysteine-containing peptides for mass spectrometry analysis
  • Provided in convenient single-use, no-weigh format
  • High-purity, crystalline reagent
  • Compatible with EasyPep chemistry and High Select Phosphopeptide Enrichment kits
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 x 3.5 mg
Molecular Weight (g/mol) 348.99
Percent Purity >90%
Grade MS grade
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.