missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetic acid, 97+%

Product Code. 11499583
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499583 100 g 100g
11489583 25 g 25g
11409593 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499583 Supplier Thermo Scientific Alfa Aesar Supplier No. A12410.22
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64-69-7 | C2H3IO2 | 185.948 g/mol

Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.

Solubility
Soluble in water, alcohol and ethanol. Slightly soluble in ether and chloroform.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents, bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-69-7
Molecular Formula C2H3IO2
Molecular Weight (g/mol) 185.948
MDL Number MFCD00002685
InChI Key JDNTWHVOXJZDSN-UHFFFAOYSA-N
Synonym iodoacetic acid, acetic acid, iodo, iodoacetate, monoiodoacetic acid, monoiodoacetate, monoiodine acetate, iodoaceticacid, iodoethanoic acid, acetic acid, 2-iodo, kyselina jodoctova
PubChem CID 5240
ChEBI CHEBI:74571
IUPAC Name 2-iodoacetic acid
SMILES C(C(=O)O)I

Specifications

Melting Point 79°C to 82°C
Boiling Point 208°C (decomposition)
Odor Pungent
Quantity 100 g
UN Number UN2923
Beilstein 1739079
Sensitivity Light sensitive
Merck Index 14,5025
Solubility Information Soluble in water,alcohol and ethanol. Slightly soluble in ether and chloroform.
Formula Weight 185.95
Percent Purity ≥97%
Chemical Name or Material Iodoacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.