missing translation for 'onlineSavingsMsg'
Learn More

Iodoacetic Acid, TRC

Product Code. 30137912
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137912 50 g 50g
30133383 10 g 10g
30041255 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137912 Supplier Toronto Research Chemicals Supplier No. TRCI68552050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-69-7
Molecular Formula C2 H3 I O2
Molecular Weight (g/mol) 185.95
Synonym 2-Iodoacetic acid (ACI), Acetic acid, iodo- (8CI, 9CI), CH2ICOOH, Iodoacetates, Iodoacetic acid, MIA, Monoiodoacetic acid, 2-Iodoacetic acid (ACI), Acetic acid, iodo- (8CI, 9CI), CH2ICOOH, Iodoacetates, Iodoacetic acid, MIA, Monoiodoacetic acid
IUPAC Name 2-iodoacetic acid
SMILES OC(=O)CI

Specifications

Quantity 50 g
InChI Formula InChI=1S/C2H3IO2/c3-1-2(4)5/h1H2,(H,4,5)
Formula Weight 185.9178
Chemical Name or Material Iodoacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.